missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopentane, tech. 90%, Thermo Scientific Chemicals

Product Code. 11413427
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chemical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-81-3
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000160
InChI Key LGAJYTCRJPCZRJ-UHFFFAOYSA-N
Synonym pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
PubChem CID 7890
IUPAC Name 2-bromopentane
SMILES CCCC(C)Br

Specifications

Melting Point -96°C
Density 1.22
Boiling Point 117°C to 118°C
Flash Point 20°C (68°F)
Odor Characteristic
Refractive Index 1.4403
Quantity 100 g
UN Number UN2343
Beilstein 1718829
Solubility Information Not miscible or difficult to mix in water. Soluble in Dichloromethane,Ether,Ethyl Acetate,Hexane and Methanol
Formula Weight 151.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Bromopentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'