missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylacetone, 99%, Thermo Scientific Chemicals

13.67 € - 45.83 €

Chemical Identifiers

CAS 21906-31-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.08
MDL Number MFCD03410431
InChI Key TZIAZLUAMDLDJF-UHFFFAOYSA-N
Synonym 2-bromophenylacetone, 1-2-bromophenyl propan-2-one, 2-bromophenyl acetone, 1-2-bromophenyl-2-propanone, 1-acetonyl-2-bromobenzene, 2-propanone, 1-2-bromophenyl, o-bromo benyl methyl ketone, 1-2-bromophenyl acetone, o-bromo benzyl methyl ketone, pubchem23388
PubChem CID 2734092
IUPAC Name 1-(2-bromophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10110132
SDS View Documents
Thermo Scientific Acros
350590010
1 g Glass bottle
13.67 €
1g
Please to purchase this item. Need a web account? Register with us today!
10068761
SDS View Documents
Thermo Scientific Acros
350590050
5 g Glass bottle
45.83 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Bromophenylacetone

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromophenylacetone, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.