Learn More
(2-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals
32.96 € - 462.00 €
Chemical Identifiers
CAS | 38274-16-7 |
---|---|
Molecular Formula | C11H13BrSi |
Molecular Weight (g/mol) | 253.21 |
MDL Number | MFCD01321391 |
InChI Key | FABNGXSCLHXUOH-UHFFFAOYSA-N |
Synonym | 2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl |
PubChem CID | 4547398 |
SMILES | C[Si](C)(C)C#CC1=CC=CC=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410178
|
Thermo Scientific Alfa Aesar
H53402.03 |
1 g |
32.96 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420178
|
Thermo Scientific Alfa Aesar
H53402.06 |
5 g |
109.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430178
|
Thermo Scientific Alfa Aesar
H53402.14 |
25 g |
462.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
38274-16-7 | |
253.21 | |
FABNGXSCLHXUOH-UHFFFAOYSA-N | |
4547398 |
C11H13BrSi | |
MFCD01321391 | |
2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl | |
C[Si](C)(C)C#CC1=CC=CC=C1Br |
Specifications
60°C to 80°C (2mmHg) | |
C11H13BrSi | |
>113°C (235°F) | |
Slightly soluble in water. | |
C[Si](C)(C)C#CC1=CC=CC=C1Br | |
253.21 | |
253.21 | |
1.554 | |
(2-Bromophenylethynyl)trimethylsilane |
38274-16-7 | |
MFCD01321391 | |
2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl | |
FABNGXSCLHXUOH-UHFFFAOYSA-N | |
[2-(2-bromophenyl)ethynyl]trimethylsilane | |
4547398 | |
98% | |
1.19 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only