missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropane, 99%, Thermo Scientific Chemicals

Product Code. 15179153
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-26-3
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.99
InChI Key NAMYKGVDVNBCFQ-UHFFFAOYSA-N
Synonym isopropyl bromide, propane, 2-bromo, isopropylbromide, 2-bromo-propane, sec-propyl bromide, unii-r651xov97z, ccris 7919, hsdb 623, i-propylbromide, 2-brompropan
PubChem CID 6358
IUPAC Name 2-bromopropane
SMILES CC(C)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -89.0°C
Color Brown to Colorless
Density 1.3100g/mL
Boiling Point 59.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Drum
Linear Formula (CH3)2CHBr
Refractive Index 1.4250 to 1.4270
Quantity 10 L
Beilstein 01,108
Fieser 09,69
Merck Index 15,5256
Specific Gravity 1.31
Solubility Information Solubility in water: 0.3g/100mL. Other solubilities: miscible with alcohol,benzene,chloroform,ether,and acetone
Formula Weight 122.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromopropane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.