missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine, 99%, Thermo Scientific Chemicals

29.75 € - 289.44 €

Chemical Identifiers

CAS 109-04-6
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 157.998
MDL Number MFCD00006219
InChI Key IMRWILPUOVGIMU-UHFFFAOYSA-N
Synonym o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8
PubChem CID 7973
ChEBI CHEBI:51574
IUPAC Name 2-bromopyridine
SMILES C1=CC=NC(=C1)Br
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11450773
View Documents
Thermo Scientific Alfa Aesar
A13241.14
25 g
29.75 €
25g
Please to purchase this item. Need a web account? Register with us today!
11460773
View Documents
Thermo Scientific Alfa Aesar
A13241.22
100 g
89.24 €
100g
Please to purchase this item. Need a web account? Register with us today!
11339659
View Documents
Thermo Scientific Alfa Aesar
A13241.30
250 g
167.84 €
250g
Please to purchase this item. Need a web account? Register with us today!
11470773
View Documents
Thermo Scientific Alfa Aesar
A13241.36
500 g
289.44 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

Solubility
Slightly miscible with water.

Notes
Keep away from heat, sparks, and flame. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and acid chlorides.
Specifications

Chemical Identifiers

109-04-6
157.998
IMRWILPUOVGIMU-UHFFFAOYSA-N
7973
2-bromopyridine
C5H4BrN
MFCD00006219
o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8
CHEBI:51574
C1=CC=NC(=C1)Br

Specifications

109-04-6
192°C to 194°C
Characteristic
1.572
UN2810
o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8
IMRWILPUOVGIMU-UHFFFAOYSA-N
2-bromopyridine
7973
158
2-Bromopyridine
1.625
97°C (207°F)
C5H4BrN
MFCD00006219
105789
Slightly miscible with water.
C1=CC=NC(=C1)Br
157.998
CHEBI:51574
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P362-P374-P380-P501c

H226-H301+H331-H310-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 203-641-6

missing translation for 'rtecsNumber' : US3850000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.