missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromothiazole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Description
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
Specifications
Specifications
Density | 1.836 |
Boiling Point | 171°C |
Flash Point | 63°C (145°F) |
Refractive Index | 1.593 |
Quantity | 250 g |
Beilstein | 105724 |
Solubility Information | It is insoluble in water. |
Formula Weight | 164.02 |
Percent Purity | 99% |
Chemical Name or Material | 2-Bromothiazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'