missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotoluene, 99%

Product Code. 10705561 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10705561

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 95-46-5 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-46-5
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000068
InChI Key QSSXJPIWXQTSIX-UHFFFAOYSA-N
Synonym 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
PubChem CID 7236
IUPAC Name 1-bromo-2-methylbenzene
SMILES CC1=CC=CC=C1Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -27°C
Color Colorless to Yellow
Density 1.4200g/mL
Boiling Point 181.5°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4Br
Refractive Index 1.5550 to 1.557
Quantity 25 mL
Beilstein 05, 304
Merck Index 14, 1439
Specific Gravity 1.42
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromotoluene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.