Learn More
2-Butanone, 99%, Thermo Scientific Chemicals
Description
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.
Solubility
Miscible with cold water, diethyl ether and acetone.
Notes
It is stable, incompatible with oxidizing agents, bases, strong reducing agents.

Specifications
Specifications
Melting Point | -87°C |
Density | 0.805 |
Boiling Point | 80°C |
Quantity | 1000 mL |
Flash Point | −3°C (26°F) |
Odor | Characteristic |
Refractive Index | 1.3785 |
Linear Formula | CH3COCH2CH3 |
UN Number | UN1193 |
Beilstein | 741880 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252891 | Certificate of Analysis | 28/10/2024 | L13185.AU,L13185.0F,L13185.AP |
T07J032 | Certificate of Analysis | 13/07/2023 | L13185.AU,L13185.0F,L13185,L13185.AP |
T07J031 | Certificate of Analysis | 13/07/2023 | L13185.AU,L13185.0F,L13185,L13185.AP |
10247297 | Certificate of Analysis | 11/07/2023 | L13185.AU,L13185.0F,L13185,L13185.AP |
10243294 | Certificate of Analysis | 07/09/2022 | L13185.AU,L13185.0F,L13185.AP |
Safety and Handling


- 2-Butanone
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Methyl ethyl ketone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.