missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, HPLC Grade, 99.5+%

Product Code. 11338197
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11338197

missing translation for 'mfr': Thermo Scientific Alfa Aesar 022924.M1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-93-3 | C4H8O | 72.11 g/mol

2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87°C
Color Colorless
Density 0.805 g/mL
Boiling Point 80°C
Quantity 4 x 1 L
Flash Point -3°C (26°F)
Odor Characteristic
Physical Form Colorless liquid
Refractive Index 1.379
Linear Formula CH3COCH2CH3
UN Number UN1193
Beilstein 741880
Merck Index 14,6072
Solubility Information Miscible with cold water,diethyl ether and acetone.
Formula Weight 72.11
Percent Purity ≥99.5%
Grade HPLC
Chemical Name or Material 2-Butanone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.