missing translation for 'onlineSavingsMsg'
Learn More
2-Butanone, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™ Product Code.: 15663950

2-Butanone, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™

Product Code. 15663950
4 x 2.5 L, Glass Bottle
Click to view available options
Quantity:
250 mL
4 x 2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15663950

Brand: Honeywell-Riedel-de Haen 334074X2.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -87°C
Density 0.805g/mL at 25°C
Boiling Point 79°C to 81°C
Quantity 4 x 2.5 L
Flash Point −3°C (26.6°F)
Odor Solvent like
Packaging Glass Bottle
Solubility Information 290g/L at 20°C
compliance-icons compliance-icons
Product Identifier
  • 2-Butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butanone, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.