missing translation for 'onlineSavingsMsg'
Learn More
2-Butanone, technical Product Code.: 13466369

2-Butanone, technical

Product Code. 13466369
5 L, Plastic Drum
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
This item is not returnable. View return policy

Product Code. 13466369

Brand: Thermo Scientific Acros 444170050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-93-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -87.0°C
CAS 78-93-3
Density 0.8060g/mL
Boiling Point 80.0°C
Quantity 5 L
Flash Point −7°C
Packaging Plastic Drum
Refractive Index 1.3780 to 1.3800
Specific Gravity 0.806
Molecular Formula C4H8O
Linear Formula C2H5COCH3
MDL Number MFCD00011648
Viscosity 0.42 mPa.s (15°C)
Merck Index 15,6143
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene,alcohol and ether,miscible with oils
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
SMILES CCC(C)=O
IUPAC Name butan-2-one
Molecular Weight (g/mol) 72.11
Formula Weight 72.11
Grade Technical
Chemical Name or Material 2-Butanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468101 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number444170050
Lot NumberA0442868 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number444170050
Lot NumberA0437085 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number444170050
Lot NumberA0461040 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number444170050
Lot NumberA0453485 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number444170050
compliance-icons compliance-icons
Product Identifier
  • 2-Butanone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Methyl ethyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butanone, technical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.