missing translation for 'onlineSavingsMsg'
Learn More
2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 10532171

2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10532171
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
This item is not returnable. View return policy

Product Code. 10532171

Brand: Thermo Scientific Acros 154330010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
PubChem CID 8133
ChEBI CHEBI:63921
SMILES CCCCOCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Density 0.9010g/mL
Boiling Point 171.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)3OCH2CH2OH
Refractive Index 1.4180 to 1.4200
Quantity 1 L
Beilstein 01, II, 519
Residue after Evaporation 0.02% max.
Merck Index 15, 1560
Specific Gravity 0.901
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvents
IUPAC Name 2-butoxyethan-1-ol
Viscosity 5.31 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Butoxyethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463029 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number154330025,154330100,154330010,154330250
Lot NumberA0456235 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number154330025,154330100,154330010,154330250
Lot NumberA0396622 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number154330025,154330100,154330010,154330250
Lot NumberA0404418 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number154330025,154330100,154330010,154330250
Lot NumberA0372594 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number154330025,154330100,154330010,154330250
compliance-icons
Product Identifier
  • 2-Butoxyethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butoxyethanol, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.