missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl-1H-imidazol-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 10082404
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 68283-19-2
Molecular Formula C8H14N2O
Molecular Weight (g/mol) 154.21
MDL Number MFCD00239516
InChI Key UZKBZGAMRJRWLR-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
PubChem CID 2735673
IUPAC Name (2-butyl-1H-imidazol-5-yl)methanol
SMILES CCCCC1=NC=C(CO)N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material (2-Butyl-1H-imidazol-4-yl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.