missing translation for 'onlineSavingsMsg'
Learn More

2-Butyrylthiophene, 97%, Thermo Scientific™

Product Code. 11427317
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5333-83-5
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00022503
InChI Key YXHIINNJOGKPLF-UHFFFAOYSA-N
Synonym 2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz
PubChem CID 79248
SMILES CCCC(=O)C1=CC=CS1

Specifications

Density 1.09
Boiling Point 242°C to 244°C
Flash Point 102°C (216°F)
Odor Characteristic
Refractive Index 1.543
Quantity 10 g
Beilstein 110167
IUPAC Name 1-thiophen-2-ylbutan-1-one
Formula Weight 154.23
Percent Purity 97%
Chemical Name or Material 2-Butyrylthiophene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'