missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butyrylthiophene, 97%, Thermo Scientific™
21.80 € - 122.00 €
Chemical Identifiers
CAS | 5333-83-5 |
---|---|
Molecular Formula | C8H10OS |
Molecular Weight (g/mol) | 154.23 |
MDL Number | MFCD00022503 |
InChI Key | YXHIINNJOGKPLF-UHFFFAOYSA-N |
Synonym | 2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz |
PubChem CID | 79248 |
SMILES | CCCC(=O)C1=CC=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309579
|
Thermo Scientific Alfa Aesar
A15048.06 |
5 g |
21.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427317
|
Thermo Scientific Alfa Aesar
A15048.09 |
10 g | |||||||
11437317
|
Thermo Scientific Alfa Aesar
A15048.18 |
50 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5333-83-5 | |
154.23 | |
YXHIINNJOGKPLF-UHFFFAOYSA-N | |
79248 |
C8H10OS | |
MFCD00022503 | |
2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz | |
CCCC(=O)C1=CC=CS1 |
Specifications
5333-83-5 | |
242°C to 244°C | |
Characteristic | |
1.543 | |
110167 | |
YXHIINNJOGKPLF-UHFFFAOYSA-N | |
1-(thiophen-2-yl)butan-1-one | |
79248 | |
97% |
1.09 | |
102°C (216°F) | |
C8H10OS | |
MFCD00022503 | |
2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz | |
CCCC(=O)C1=CC=CS1 | |
154.23 | |
154.23 | |
2-Butyrylthiophene |
Safety and Handling
missing translation for 'einecsNumber' : 226-246-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures