missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium chloride, 90%, Thermo Scientific Chemicals

Product Code. 11946271
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37091-73-9
Molecular Formula C5H10Cl2N2
Molecular Weight (g/mol) 169.05
MDL Number MFCD09039290
InChI Key AEBBXVHGVADBHA-UHFFFAOYSA-M
Synonym 2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
PubChem CID 10176306
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
SMILES [Cl-].CN1CC[N+](C)=C1Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 112°C to 121°C
Color White
Elemental Analysis 31.26 to 39.79% C, 14.58 to 18.56% N
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15,3434
Formula Weight 169.05
Percent Purity 90%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolinium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-1,3-dimethylimidazolinium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'