missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,3-dinitrobenzene, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.
Solubility
Soluble in alcohol. Moderately soluble in benzene, ether. Practically insoluble in water.
Notes
Incompatible materials are oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.
Solubility
Soluble in alcohol. Moderately soluble in benzene, ether. Practically insoluble in water.
Notes
Incompatible materials are oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Melting Point | 83°C to 88°C |
Density | 1.687 |
Boiling Point | 315°C |
Quantity | 1 g |
UN Number | UN3441 |
Beilstein | 522187 |
Merck Index | 14,2137 |
Solubility Information | Soluble in alcohol. Moderately soluble in benzene,ether. Practically insoluble in water. |
Formula Weight | 202.55 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'