missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylimidazole, 97+%, Thermo Scientific Chemicals

76.40 € - 215.00 €

Chemical Identifiers

CAS 253453-91-7
Molecular Formula C4H5ClN2
Molecular Weight (g/mol) 116.55
MDL Number MFCD02179531
InChI Key VSCQFRRKWFQPOA-UHFFFAOYSA-N
PubChem CID 2773332
IUPAC Name 2-chloro-1-methylimidazole
SMILES CN1C=CN=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15438357
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51867.MD
250 mg
76.40 €
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15428357
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51867.03
1 g
215.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

253453-91-7
116.55
VSCQFRRKWFQPOA-UHFFFAOYSA-N
2-chloro-1-methylimidazole
C4H5ClN2
MFCD02179531
2773332
CN1C=CN=C1Cl

Specifications

253453-91-7
C4H5ClN2
2-chloro-1-methyl-1h-imidazole, 1h-imidazole, 2-chloro-1-methyl, pubchem7596, acmc-209gjb, 1-methyl-2-chloro-1h-imidazole, 2-chloro-n-methylimidazole, 1h-imidazole,2-chloro-1-methyl, 2-chloro-1n-methyl-imidazole, 2-chloro-1h-imidazole, n-me, 2-chloro-1h-imidazole, n-methyl
CN1C=CN=C1Cl
116.55
116.55
2-Chloro-1-methylimidazole
-10°C
MFCD02179531
VSCQFRRKWFQPOA-UHFFFAOYSA-N
2-chloro-1-methylimidazole
2773332
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only