missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylbutane, 95%, Thermo Scientific Chemicals

Product Code. 11469213
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-36-5
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000818
InChI Key CRNIHJHMEQZAAS-UHFFFAOYSA-N
Synonym tert-amyl chloride, butane, 2-chloro-2-methyl, 2-methyl-2-chlorobutane, tert-pentyl chloride, 1,1-dimethylpropyl chloride, tertiary pentyl chloride, t-pentyl chloride, acmc-209mdc, 2-chloro-2-methyl butane, 2-chloro-2-methyl-butane
PubChem CID 61143
IUPAC Name 2-chloro-2-methylbutane
SMILES CCC(C)(C)Cl

Specifications

Melting Point -73°C
Density 0.866
Boiling Point 85°C to 86°C
Flash Point −10°C (14°F)
Refractive Index 1.405
Quantity 5 g
UN Number UN1107
Beilstein 1696930
Solubility Information Soluble in ethanol,ethyl ether. Insoluble in water.
Formula Weight 106.6
Percent Purity 95%
Chemical Name or Material 2-Chloro-2-methylbutane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-2-methylbutane, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.