missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dimethylpyrazine, 97%, Thermo Scientific™

Product Code. 10397663
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 38557-72-1
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00126945
InChI Key BTGGHNHGPURMEO-UHFFFAOYSA-N
PubChem CID 11672680
IUPAC Name 2-chloro-3,5-dimethylpyrazine
SMILES CC1=CN=C(Cl)C(C)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-chloro 3,5-dimethyl pyarazine, 2-chloro-3,5-dimethyl-1,4-diazine, 2-chloro-3,5-dimethyl-pyrazine, 3,5-dimethyl-2-chloropyrazine, 2-chloro 3,5-dimethyl pyrazine, 2-chloro-3,5-dimethyl pyrazine, pyrazine,2-chloro-3,5-dimethyl, 2-chloranyl-3,5-dimethyl-pyrazine, pyrazine, 2-chloro-3,5-dimethyl, 3-chloro-2,6-dimethylpyrazine
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-3, 5-dimethylpyrazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'