missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylthiophene, 97%, Thermo Scientific™

Product Code. 11317549
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14345-97-2
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.61
MDL Number MFCD00130083
InChI Key KQFADYXPELMVHE-UHFFFAOYSA-N
Synonym 2-chlor-3-methylthiophen, thiophene, 2-chloro-3-methyl, pubchem5164, acmc-209cqk, 2-chloro 3-methyl thiophene, 2-chloro-3-methyl thiophene, ksc174m0h, 2-chloro-3-methylthiophene
PubChem CID 84365
IUPAC Name 2-chloro-3-methylthiophene
SMILES CC1=C(Cl)SC=C1

Specifications

Density 1.211
Boiling Point 147°C to 149°C
Flash Point 46°C (115°F)
Refractive Index 1.54
Quantity 5 g
UN Number UN1993
Formula Weight 132.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-methylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'