missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-4',5'-difluoroacetophenone, 95%, Thermo Scientific™

Product Code. 11385315
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121872-94-4
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD00671761
InChI Key BNODNMUCMFITCS-UHFFFAOYSA-N
Synonym 2'-chloro-4',5'-difluoroacetophenone, 1-2-chloro-4,5-difluorophenyl ethanone, 2-chloro-4,5-difluoroacetophenone, 1-2-chloro-4,5-difluoro-phenyl ethanone, ethanone,1-2-chloro-4,5-difluorophenyl, ethanone, 1-2-chloro-4,5-difluorophenyl, 2-chloro-4 ,5-difluoroacetophenone, 1-acetyl-2-chloro-4,5-difluorobenzene, acmc-20dywg, pubchem4109
PubChem CID 2736489
IUPAC Name 1-(2-chloro-4,5-difluorophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1Cl)F)F

Specifications

Quantity 1 g
Formula Weight 190.58
Percent Purity 95%
Chemical Name or Material 2'-Chloro-4',5'-difluoroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'