Learn More
2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine, 96%, Thermo Scientific™
43.00 € - 125.00 €
Chemical Identifiers
CAS | 122-34-9 |
---|---|
Molecular Formula | C7H12ClN5 |
Molecular Weight (g/mol) | 201.658 |
MDL Number | MFCD00023174 |
InChI Key | ODCWYMIRDDJXKW-UHFFFAOYSA-N |
Synonym | simazine, gesatop, princep, simanex, aquazine, batazina, herbazin, symazine, tafazine, taphazine |
PubChem CID | 5216 |
ChEBI | CHEBI:27496 |
IUPAC Name | 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine |
SMILES | CCNC1=NC(=NC(=N1)Cl)NCC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
122-34-9 | |
201.658 | |
ODCWYMIRDDJXKW-UHFFFAOYSA-N | |
5216 | |
6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine |
C7H12ClN5 | |
MFCD00023174 | |
simazine, gesatop, princep, simanex, aquazine, batazina, herbazin, symazine, tafazine, taphazine | |
CHEBI:27496 | |
CCNC1=NC(=NC(=N1)Cl)NCC |
Specifications
122-34-9 | |
C7H12ClN5 | |
UN3077 | |
14,8533 | |
ODCWYMIRDDJXKW-UHFFFAOYSA-N | |
6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine | |
5216 | |
201.66 | |
2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine |
227°C to 229°C | |
MFCD00023174 | |
10895 | |
simazine, gesatop, princep, simanex, aquazine, batazina, herbazin, symazine, tafazine, taphazine | |
CCNC1=NC(=NC(=N1)Cl)NCC | |
201.658 | |
CHEBI:27496 | |
96% |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 204-535-2
missing translation for 'rtecsNumber' : XY5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only