missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-5-nitrotoluene, 97%, Thermo Scientific Chemicals

Product Code. 15459838
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-4-fluoro-5-nitrotoluene, is used as a pharmaceutical intermediate. It is also used as Ceritinib intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-fluoro-5-nitrotoluene, is used as a pharmaceutical intermediate. It is also used as Ceritinib intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112108-73-3
Molecular Formula C7H5ClFNO2
Molecular Weight (g/mol) 189.57
MDL Number MFCD11110549
InChI Key YXVJHZWHPLOEAP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-2-methyl-4-nitro-benzene, benzene, 1-chloro-5-fluoro-2-methyl-4-nitro, acmc-2099cn, ksc682i1p, 2-chloro-4fluoro-5-nitrotoluene, 1-chloranyl-5-fluoranyl-2-methyl-4-nitro-benzene
PubChem CID 10103979
IUPAC Name 1-chloro-5-fluoro-2-methyl-4-nitrobenzene
SMILES CC1=CC(=C(C=C1Cl)F)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 189.57
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Chloro-4-fluoro-5-nitrotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'