missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 15434687
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21971-21-1
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00085943
InChI Key IBANGHTVBPZCHF-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-benzoic acid, 2-chloro-4-methyloxy benzoic acid, benzoic acid, 2-chloro-4-methoxy, 2-chloro-4-methoxybenzoicacid, 2-chloranissaure, pubchem14030, 2-chloro-p-anisic acid, ksc494q0b, 4-methoxy-2-chloro-benzoic acid
PubChem CID 12648026
IUPAC Name 2-chloro-4-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)Cl

Specifications

Quantity 5 g
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'