missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

Product Code. 11474933
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39885-50-2
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00042563
InChI Key MBBUTABXEITVNY-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
PubChem CID 162001
IUPAC Name 2-chloro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Cl)C(F)(F)F

Specifications

Density 1.434
Boiling Point 214°C to 218°C
Flash Point 103°C (217°F)
Refractive Index 1.5015
Quantity 5 g
UN Number UN2810
Beilstein 4179703
Formula Weight 195.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-4- (trifluoromethyl)aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'