missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(difluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15448015
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-54-2
Molecular Formula C8H6BrClF2O
Molecular Weight (g/mol) 271.486
MDL Number MFCD09832368
InChI Key NLCSHYHRPBCBDK-UHFFFAOYSA-N
Synonym 2-chloro-5-difluoromethoxy benzyl bromide, 2-bromomethyl-1-chloro-4-difluoromethoxy benzene, 2-chloro-5-difluoromethoxy benzyl bromide, jrd
PubChem CID 46737682
IUPAC Name 2-(bromomethyl)-1-chloro-4-(difluoromethoxy)benzene
SMILES C1=CC(=C(C=C1OC(F)F)CBr)Cl

Specifications

Refractive Index 1.535
Quantity 250 mg
UN Number UN3265
Formula Weight 271.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(difluoromethoxy)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'