missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-fluoropyrimidine, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Beschreibung
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
Notes
Incompatible with strong oxidizing agents.
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
Notes
Incompatible with strong oxidizing agents.
Spezifikation
Spezifikation
Density | 1.073 |
Flash Point | 65°C (149°F) |
Refractive Index | 1.503 |
Quantity | 5 g |
UN Number | UN3267 |
Formula Weight | 132.52 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-5-fluoropyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'