missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-methylphenol, 99%, Thermo Scientific Chemicals

50.20 € - 542.70 €

Chemical Identifiers

CAS 615-74-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00002175
InChI Key SMFHPCZZAAMJJO-UHFFFAOYSA-N
Synonym 6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
PubChem CID 12008
IUPAC Name 2-chloro-5-methylphenol
SMILES CC1=CC=C(Cl)C(O)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10625422
View Documents
Thermo Scientific Acros
170410250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10573341
View Documents
Thermo Scientific Acros
170411000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10455755
View Documents
Thermo Scientific Acros
170415000
500 g Glass bottle
542.70 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

615-74-7
142.58
SMFHPCZZAAMJJO-UHFFFAOYSA-N
12008
CC1=CC=C(Cl)C(O)=C1
C7H7ClO
MFCD00002175
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
2-chloro-5-methylphenol

Specifications

615-74-7
100.0
Beige
81°C
98.5% min. (GC)
ClC6H3(CH3)OH
06, I, 187
SMFHPCZZAAMJJO-UHFFFAOYSA-N
2-chloro-5-methylphenol
12008
99%
2-chloro-5-methylphenol
98.5
45.0°C to 48.0°C
196.0°C
Authentic
C7H7ClO
MFCD00002175
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
CC1=CC=C(Cl)C(O)=C1
142.58
142.58
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-444-9

Product Identifier
  • 2-Chloro-5-methylphenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only