missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-n-decylpyrimidine, 99%, Thermo Scientific™

Product Code. 11444957
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170434-06-7
Molecular Formula C14H23ClN2
Molecular Weight (g/mol) 254.80
MDL Number MFCD02683077
InChI Key LJRSECCCAXFYFU-UHFFFAOYSA-N
Synonym 2-chloro-5-n-decylpyrimidine
PubChem CID 22099083
IUPAC Name 2-chloro-5-decylpyrimidine
SMILES CCCCCCCCCCC1=CN=C(Cl)N=C1

Specifications

Quantity 1 g
Formula Weight 254.8
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-n-decylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'