Learn More
2-chloro-5-nitropyridine, 99%, Thermo Scientific Chemicals
99.29 € - 276.38 €
Chemical Identifiers
CAS | 4548-45-2 |
---|---|
Molecular Formula | C5H3ClN2O2 |
Molecular Weight (g/mol) | 158.54 |
MDL Number | MFCD00006240 |
InChI Key | BAZVFQBTJPBRTJ-UHFFFAOYSA-N |
Synonym | pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin |
PubChem CID | 78308 |
IUPAC Name | 2-chloro-5-nitropyridine |
SMILES | [O-][N+](=O)C1=CC=C(Cl)N=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10665332
|
Thermo Scientific Acros
109740250 |
25 g | Glass bottle |
99.29 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666934
|
Thermo Scientific Acros
109741000 |
100 g | Glass bottle |
276.38 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4548-45-2 | |
158.54 | |
BAZVFQBTJPBRTJ-UHFFFAOYSA-N | |
78308 | |
[O-][N+](=O)C1=CC=C(Cl)N=C1 |
C5H3ClN2O2 | |
MFCD00006240 | |
pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin | |
2-chloro-5-nitropyridine |
Specifications
4548-45-2 | |
100.0 | |
Yellow to Beige | |
98.5% min. (GC) | |
Cl(C5H3N)NO2 | |
20, I, 82 | |
Solubility in water: insoluble | |
[O-][N+](=O)C1=CC=C(Cl)N=C1 | |
158.54 | |
158.54 | |
Crystalline Powder |
98.5 | |
106°C to 110°C | |
Authentic | |
C5H3ClN2O2 | |
MFCD00006240 | |
pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin | |
BAZVFQBTJPBRTJ-UHFFFAOYSA-N | |
2-chloro-5-nitropyridine | |
78308 | |
99% | |
2-Chloro-5-nitropyridine |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 224-908-3
- 2-Chloro-5-nitropyridine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.