missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-5'-(trifluoromethyl)acetophenone, 96%, Thermo Scientific Chemicals

Product Code. 11415537
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2'-Chloro-5'-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Chloro-5′-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.11g/L @ 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71648-45-8
Molecular Formula C9H6ClF3O
Molecular Weight (g/mol) 222.591
MDL Number MFCD00045092
InChI Key YRGBMTWHOFQSDJ-UHFFFAOYSA-N
Synonym 2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210
PubChem CID 144502
IUPAC Name 1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl

Specifications

Quantity 1 g
Solubility Information Sparingly soluble in water 0.11g/L @ 25°C.
Formula Weight 222.6
Percent Purity ≥97%
Chemical Name or Material 2'-Chloro-5'-(trifluoromethyl)acetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'