missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

47.00 € - 530.00 €

Chemical Identifiers

CAS 505084-59-3
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD08741353
InChI Key RMSKYTKOFDQVEL-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl nicotinic acid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-5-trifluoromethyl-3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethylnicotinic acid, 2-chloro-5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, ksc269c4j, 2-chloro-5-trifluoromethyl nicotinicacid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic, 2-chloro-5-trifluoromethyl pyridine-3-carboxylicacid
PubChem CID 12140227
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=C(C(=NC=C1C(F)(F)F)Cl)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428266
View Documents
Thermo Scientific Alfa Aesar
H63350.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15499998
View Documents
Thermo Scientific Alfa Aesar
H63350.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15400009
View Documents
Thermo Scientific Alfa Aesar
H63350.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

505084-59-3
225.551
RMSKYTKOFDQVEL-UHFFFAOYSA-N
12140227
C1=C(C(=NC=C1C(F)(F)F)Cl)C(=O)O
C7H3ClF3NO2
MFCD08741353
2-chloro-5-trifluoromethyl nicotinic acid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-5-trifluoromethyl-3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethylnicotinic acid, 2-chloro-5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, ksc269c4j, 2-chloro-5-trifluoromethyl nicotinicacid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic, 2-chloro-5-trifluoromethyl pyridine-3-carboxylicacid
2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid

Specifications

505084-59-3
C7H3ClF3NO2
2-chloro-5-trifluoromethyl nicotinic acid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-5-trifluoromethyl-3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethylnicotinic acid, 2-chloro-5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, ksc269c4j, 2-chloro-5-trifluoromethyl nicotinicacid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic, 2-chloro-5-trifluoromethyl pyridine-3-carboxylicacid
C1=C(C(=NC=C1C(F)(F)F)Cl)C(=O)O
225.551
225.55
Solid
173°C to 178°C
MFCD08741353
RMSKYTKOFDQVEL-UHFFFAOYSA-N
2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
12140227
97%
2-Chloro-5-(trifluoromethyl)nicotinic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.