missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-6'-fluoroacetophenone, 97%, Thermo Scientific™

Product Code. 11461057
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Chloro-6′-fluoroacetophenone is used in medicine & organic intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87327-69-3
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD01631390
InChI Key DNVGZKIRMBCQEQ-UHFFFAOYSA-N
PubChem CID 2773578
SMILES CC(=O)C1=C(F)C=CC=C1Cl

Specifications

Refractive Index 1.511
Quantity 5 g
Beilstein 5862530
Synonym 2'-chloro-6'-fluoroacetophenone, 1-2-chloro-6-fluorophenyl ethanone, 2-chloro-6-fluoroacetophenone, 1-2-chloro-6-fluorophenyl ethan-1-one, ethanone, 1-2-chloro-6-fluorophenyl, pubchem7110, 1-acetyl-2-chloro-6-fluorobenzene, 1-acetyl-6-chloro-2-fluorobenzene, 1-2-chloro-6-fluoro-phenyl-ethanone, ethanone,1-2-chloro-6-fluorophenyl
Solubility Information Sparingly soluble in water.
IUPAC Name 1-(2-chloro-6-fluorophenyl)ethan-1-one
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 2'-Chloro-6'-fluoroacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.