missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-methyl-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10634965
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 22123-14-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00052900
InChI Key SXLBWNSGCIEART-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, pyridine, 2-chloro-6-methyl-4-trifluoromethyl, pubchem9219, 6-chloro-4-trifluoromethyl-2-picoline, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine
PubChem CID 2736626
IUPAC Name 2-chloro-6-methyl-4-(trifluoromethyl)pyridine
SMILES CC1=NC(Cl)=CC(=C1)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 50 g
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methyl-4-(trifluoromethyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'