missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylnicotinamide, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 170.6 |
Percent Purity | 95% |
Chemical Name or Material | 2-Chloro-6-methylnicotinamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Chloro-6-methylnicotinamide, ≥95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction