missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11456907
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson's reagent and solvent tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson′s reagent and solvent tetrahydrofuran.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-32-1
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00045170
InChI Key WGNNILPYHCKCFF-UHFFFAOYSA-N
Synonym n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline
PubChem CID 136736
IUPAC Name 2-chloro-N-methylaniline
SMILES CNC1=CC=CC=C1Cl

Specifications

Density 1.15
Boiling Point 218°C
Flash Point 92°C (197°F)
Refractive Index 1.58
Quantity 25 g
UN Number UN2810
Beilstein 2802444
Solubility Information Not miscible or difficult to mix.
Formula Weight 141.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'