missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10028691
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-23-9
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051520
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
PubChem CID 76187
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange-Red to Yellow
Density 1.5480g/mL
Boiling Point 150.0°C to 152.0°C (12.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4SO2Cl
Refractive Index 1.5745 to 1.5765
Quantity 1 g
Specific Gravity 1.548
IUPAC Name 2-chlorobenzene-1-sulfonyl chloride
Formula Weight 211.07
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Chlorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.