missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl mercaptan, 98%, Thermo Scientific™

Product Code. 10455433 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10455433

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 39718-00-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.65
MDL Number MFCD00004868
InChI Key WWFIIZLHSNBNTC-UHFFFAOYSA-N
Synonym 2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw
PubChem CID 580759
IUPAC Name (2-chlorophenyl)methanethiol
SMILES C1=CC=C(C(=C1)CS)Cl

Specifications

CAS Min % 97.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.2200g/mL
Boiling Point 120°C to 121°C (25.0 mmHg)
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5890 to 1.591
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.22
Linear Formula ClC6H4CH2SH
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzyl mercaptan
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorobenzyl mercaptan
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.