missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroisonicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10496273
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy
2-Chloroisonicotinaldehyde, ≥97%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101066-61-9
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.55
MDL Number MFCD06651557
InChI Key UFPOSTQMFOYHJI-UHFFFAOYSA-N
Synonym 2-chloroisonicotinaldehyde, 2-chloro-4-pyridinecarboxaldehyde, 2-chloro-4-formylpyridine, 2-chloro-pyridine-4-carbaldehyde, 2-chloropyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde, 2-chloro, pubchem5099, 6-chloroisonicotinaldehyde, ksc180g9n, 2-chloropyridin-4-carbaldehyde
PubChem CID 2762994
IUPAC Name 2-chloropyridine-4-carbaldehyde
SMILES ClC1=CC(C=O)=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 141.56
Percent Purity 97%
Chemical Name or Material 2-Chloroisonicotinaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'