missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroisonicotinamide, 95%, Thermo Scientific™

Product Code. 10355012
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 100859-84-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00221401
InChI Key DEMJOLRJLACBRX-UHFFFAOYSA-N
PubChem CID 2800022
IUPAC Name 2-chloropyridine-4-carboxamide
SMILES C1=CN=C(C=C1C(=O)N)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 2-chloroisonicotinamide, 4-pyridinecarboxamide, 2-chloro, 2-chloro-isonicotinamide, pubchem16074, acmc-1bpcq, 2-chloro-4-pyridinecarboxamide, 4-pyridinecarboxamide,2-chloro
Formula Weight 156.57
Percent Purity 97%
Chemical Name or Material 2-Chloroisonicotinamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'