Learn More
2-(Chloromethyl)pyridine hydrochloride, 98%, Thermo Scientific Chemicals
43.32 € - 356.78 €
Chemical Identifiers
CAS | 6959-47-3 |
---|---|
Molecular Formula | C6H7Cl2N |
Molecular Weight (g/mol) | 164.029 |
MDL Number | MFCD00012811 |
InChI Key | JPMRGPPMXHGKRO-UHFFFAOYSA-N |
Synonym | 2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0 |
PubChem CID | 23392 |
ChEBI | CHEBI:76600 |
IUPAC Name | 2-(chloromethyl)pyridine;hydrochloride |
SMILES | C1=CC=NC(=C1)CCl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483913
|
Thermo Scientific Alfa Aesar
A10226.06 |
5 g |
43.32 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493913
|
Thermo Scientific Alfa Aesar
A10226.14 |
25 g |
113.57 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403923
|
Thermo Scientific Alfa Aesar
A10226.22 |
100 g |
356.78 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.
Solubility
Soluble in water, acetone (10-50mg/ml), 95%ethanol (100mg/ml).
Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
6959-47-3 | |
164.029 | |
JPMRGPPMXHGKRO-UHFFFAOYSA-N | |
23392 | |
2-(chloromethyl)pyridine;hydrochloride |
C6H7Cl2N | |
MFCD00012811 | |
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0 | |
CHEBI:76600 | |
C1=CC=NC(=C1)CCl.Cl |
Specifications
6959-47-3 | |
C6H7Cl2N | |
UN3261 | |
Hygroscopic | |
Soluble in water,acetone (10-50mg/ml),95%ethanol (100mg/ml). | |
C1=CC=NC(=C1)CCl.Cl | |
164.029 | |
CHEBI:76600 | |
98% |
120°C to 124°C | |
MFCD00012811 | |
3689155 | |
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0 | |
JPMRGPPMXHGKRO-UHFFFAOYSA-N | |
2-(chloromethyl)pyridine;hydrochloride | |
23392 | |
164.04 | |
2- (Chloromethyl)pyridine hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 230-149-9
missing translation for 'rtecsNumber' : US6825000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.