missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinamide, 98+%, Thermo Scientific™

Product Code. 11400013
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10366-35-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00006237
InChI Key ZQZAHPFFZWEUCL-UHFFFAOYSA-N
Synonym 2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
PubChem CID 82588
IUPAC Name 2-chloropyridine-3-carboxamide
SMILES C1=CC(=C(N=C1)Cl)C(=O)N

Specifications

Melting Point 163°C to 167°C
Quantity 25 g
Beilstein 119026
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloronicotinamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'