missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenol, 99%, Thermo Scientific Chemicals

Product Code. 11474063
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-57-8
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.555
MDL Number MFCD00002159
InChI Key ISPYQTSUDJAMAB-UHFFFAOYSA-N
Synonym o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
PubChem CID 7245
ChEBI CHEBI:47083
IUPAC Name 2-chlorophenol
SMILES C1=CC=C(C(=C1)O)Cl

Specifications

Melting Point 7°C to 8°C
Density 1.26
Boiling Point 175°C to 176°C
Flash Point 63°C (145°F)
Odor Phenol-like
Refractive Index 1.559
Quantity 250 g
UN Number UN2021
Beilstein 1905114
Merck Index 14,2154
Formula Weight 128.56
Percent Purity 99%
Chemical Name or Material 2-Chlorophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'