missing translation for 'onlineSavingsMsg'
Learn More

2-chlorophenylacetylene, 98%, Thermo Scientific Chemicals

Product Code. 10656620
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-31-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.58
MDL Number MFCD00269951
InChI Key DGLHLIWXYSGYBI-UHFFFAOYSA-N
Synonym 2-chlorophenylacetylene, 2'-chlorophenyl acetylene, 1-chloro-2-ethynyl-benzene, benzene, 1-chloro-2-ethynyl, 1-chlor-2-ethinylbenzol, o-chlorphenylacetylen, o-chlorophenylacetylene, pubchem2550, 2-chlorophenyl acetylene, 2;-chlorophenylacetylene
PubChem CID 585995
IUPAC Name 1-chloro-2-ethynylbenzene
SMILES C#CC1=CC=CC=C1Cl

Specifications

CAS Min % 97.5
Color Colorless to Yellow
CAS Max % 100.0
Boiling Point 82°C (16.0 hPa)
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 97.5% min. (GC)
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 2-chlorophenylacetylene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorophenylacetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'