Learn More
2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals
32.06 € - 317.58 €
Chemical Identifiers
CAS | 7623-09-8 |
---|---|
Molecular Formula | C3H4Cl2O |
Molecular Weight (g/mol) | 126.96 |
MDL Number | MFCD00000862 |
InChI Key | JEQDSBVHLKBEIZ-UHFFFAOYNA-N |
Synonym | 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone |
PubChem CID | 111019 |
IUPAC Name | 2-chloropropanoyl chloride |
SMILES | CC(C(=O)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445967
|
Thermo Scientific Alfa Aesar
B24503.14 |
25 g |
32.06 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450318
|
Thermo Scientific Alfa Aesar
B24503.22 |
100 g |
67.44 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460318
|
Thermo Scientific Alfa Aesar
B24503.36 |
500 g |
317.58 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.
Solubility
Reacts with water.
Notes
Store away from oxidizing agents, active metals, bases, water/ moisture.
Chemical Identifiers
7623-09-8 | |
126.96 | |
JEQDSBVHLKBEIZ-UHFFFAOYNA-N | |
111019 | |
CC(C(=O)Cl)Cl |
C3H4Cl2O | |
MFCD00000862 | |
2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone | |
2-chloropropanoyl chloride |
Specifications
7623-09-8 | |
109°C to 111°C | |
C3H4Cl2O | |
MFCD00000862 | |
2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone | |
JEQDSBVHLKBEIZ-UHFFFAOYNA-N | |
2-chloropropanoyl chloride | |
111019 | |
96% |
1.308 | |
31°C (88°F) | |
1.44 | |
UN2920 | |
Reacts with water. | |
CC(C(=O)Cl)Cl | |
126.96 | |
126.97 | |
2-Chloropropionyl chloride |
Safety and Handling
GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only