missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine-4-carboxamide, 98+%, Thermo Scientific™

Product Code. 11424340
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100859-84-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00221401
InChI Key DEMJOLRJLACBRX-UHFFFAOYSA-N
Synonym 2-chloroisonicotinamide, 4-pyridinecarboxamide, 2-chloro, 2-chloro-isonicotinamide, pubchem16074, acmc-1bpcq, 2-chloro-4-pyridinecarboxamide, 4-pyridinecarboxamide,2-chloro
PubChem CID 2800022
IUPAC Name 2-chloropyridine-4-carboxamide
SMILES C1=CN=C(C=C1C(=O)N)Cl

Specifications

Melting Point 201°C to 203°C
Quantity 5 g
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloropyridine-4-carboxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.