missing translation for 'onlineSavingsMsg'
Learn More
2-Chlorotoluene, 98%, Thermo Scientific Chemicals Product Code.: 11379629

2-Chlorotoluene, 98%, Thermo Scientific Chemicals

Product Code. 11379629
100 g
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11379629

Brand: Thermo Scientific Alfa Aesar B23596.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-49-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000562
InChI Key IBSQPLPBRSHTTG-UHFFFAOYSA-N
Synonym 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzeneShow MoreShow Less
PubChem CID 7238
IUPAC Name 1-chloro-2-methylbenzene
SMILES CC1=CC=CC=C1Cl

Specifications

Melting Point -36°C
Density 1.082
Boiling Point 157°C to 159°C
Quantity 100 g
Refractive Index 1.525
UN Number UN2238
Beilstein 1904175
Merck Index 14,2171
Solubility Information Slightly soluble in water.
Formula Weight 126.59
Percent Purity 99%
Chemical Name or Material 2-Chlorotoluene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberF17Z047 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberB23596.22,B23596.30,B23596.0B
Lot Number10253317 Certificate TypeCertificate of Analysis Date05/12/2024 Catalog NumberB23596.22,B23596.30,B23596.0B
Lot Number10245378 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog NumberB23596.22,B23596.30,B23596.0B
Lot Number10243503 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog NumberB23596.22,B23596.30,B23596.0B
Lot NumberF02U011 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23596.22,B23596.30,B23596.0B
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chlorotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H361d-Suspected of damaging the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : o-Chlorotoluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chlorotoluene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.