Learn More
Cytiva 2 CHR Cellulose Blotting Paper
Smooth surface. Particularly recommended for optical or radiometric scanning
Brand:  Cytiva 3002-911
Product Code. 11358854
  • 610.00 € / Pack of 100
Estimated Shipment: 11-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thickness 0.18mm. Flow rate 115mm/30 min. Slower than 1 Chr for higher resolution applications. Smooth surface. Particularly recommended for optical or radiometric scanning.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Grade 2 Chr cellulose chromatography sheets is a 0.18 mm paper for higher resolution applications.
58 x 60 cm (22.8 x 23.6 in.)
Grade 2 Chr
Cellulose
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva 2 CHR Cellulose Blotting Paper > Grade: 2CHR