missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-fluoropyridine, 98+%, Thermo Scientific Chemicals

Product Code. 11485730
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327056-62-2
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD08741371
InChI Key BHXHRMVSUUPOLX-UHFFFAOYSA-N
Synonym 2-cyano-5-fluoropyridine, 5-fluoropicolinonitrile, 5-fluoro-2-pyridinecarbonitrile, 2-cyano-5-fluorpyridine, 2-pyridinecarbonitrile, 5-fluoro, 5-fluoro-2-cyanopyridine, pubchem16479, acmc-1ag1d, 2-cyano-5-fluoro pyridine, ksc497k5h
PubChem CID 11332453
IUPAC Name 5-fluoropyridine-2-carbonitrile
SMILES C1=CC(=NC=C1F)C#N

Specifications

Melting Point 41°C to 44°C
Quantity 1 g
Formula Weight 122.1
Percent Purity ≥98%
Chemical Name or Material 2-Cyano-5-fluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'